Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CHST6 Monoclonal Antibody (OTI1D1)
GREENER_CHOICE

Catalog No. PIMA526461
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA526461 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA526461 Supplier Invitrogen™ Supplier No. MA526461
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

CHST6 Monoclonal Antibody for Western Blot

Sulfotransferase that utilizes 3'-phospho-5'-adenylyl sulfate (PAPS) as sulfonate donor to catalyze the transfer of sulfate to position 6 of non-reducing N-acetylglucosamine (GlcNAc) residues of keratan. Mediates sulfation of keratan in cornea. Keratan sulfate plays a central role in maintaining corneal transparency. Acts on the non-reducing terminal GlcNAc of short and long carbohydrate substrates that have poly-N- acetyllactosamine structures.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CHST6
Applications Western Blot
Classification Monoclonal
Clone OTI1D1
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Gene CHST6
Gene Accession No. Q9GZX3
Gene Alias carbohydrate (N-acetylglucosamine 6-O) sulfotransferase 6; carbohydrate sulfotransferase 5; carbohydrate sulfotransferase 6; carbohydrate sulfotransferase 6; C-GlcNAc6ST; CHST6; corneal N-acetylglucosamine 6-sulfotransferase; corneal N-acetylglucosamine-6-O-sulfotransferase; Galactose/N-acetylglucosamine/N-acetylglucosamine 6-O-sulfotransferase 4-beta; glcNAc6ST-5; gn6st-5; GST4-beta; hCGn6ST; LOC489707; MCDC1; N-acetylglucosamine 6-O-sulfotransferase 5
Gene Symbols CHST6, LOC489707
Host Species Mouse
Immunogen Human recombinant protein fragment corresponding to amino acids 202-395 of CHST6 produced in E.coli.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4166, 489707
Target Species Human, Mouse, Canine, Monkey
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.