Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CHST8 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA543412
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA543412 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA543412 Supplier Invitrogen™ Supplier No. PA543412
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: GCSNWKRVLM VLAGLASSTA DIQHNTVHYG SALKRLDTFD RQGILHRLST Sequence homology: Cow: 86%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 86%; Rat: 100%.

The protein encoded by this gene belongs to the sulfotransferase 2 family. It is predominantly expressed in the pituitary gland, and is localized to the Golgi membrane. This protein catalyzes the transfer of sulfate to position 4 of non-reducing N-acetylgalactosamine (GalNAc) residues in both N-glycans and O-glycans. It is responsible for sulfation of GalNAc on luteinizing hormone (LH), which is required for production of the sex hormones. Mice lacking this enzyme, exhibit increased levels of circulating LH, and precocious sexual maturation of both male and female mice. Alternatively spliced transcript variants have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CHST8
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene CHST8
Gene Accession No. Q9H2A9
Gene Alias 1500011J21Rik; AI426009; carbohydrate (N-acetylgalactosamine 4-0) sulfotransferase 8; carbohydrate N-acetylgalactosamine 4-0 sulfotransferase 8; carbohydrate sulfotransferase 8; Chst8; GalNAc-4-O-sulfotransferase 1; GalNAc4ST; GALNAC4ST1; galNAc-4-ST1; galNAc4ST-1; N-acetylgalactosamine-4-O-sulfotransferase 1; PSS3
Gene Symbols CHST8
Host Species Rabbit
Immunogen Synthetic peptide directed towards the middle region of human CHST8.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 64377
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.