Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CHST9 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5113021
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5113021 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5113021 Supplier Invitrogen™ Supplier No. PA5113021
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

CHST9 Polyclonal Antibody for Western Blot, IHC (P), ELISA

Sulfate groups in carbohydrates confer highly specific functions on glycoproteins, glycolipids, and proteoglycans and are critical for cell-cell interaction, signal transduction, and embryonic development. Sulfotransferases, such as CHST9, carry out sulfation of carbohydrates (Hiraoka et al., 2001 ).
TRUSTED_SUSTAINABILITY

Specifications

Antigen CHST9
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 7 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
Gene CHST9
Gene Accession No. Q76EC5, Q7L1S5
Gene Alias 5430438D01Rik; carbohydrate (N-acetylgalactosamine 4-0) sulfotransferase 9; carbohydrate sulfotransferase 9; CHST9; GalNAc-4-O-sulfotransferase 2; GalNAc4ST2; galNAc-4-ST2; GalNAc4ST-2; GalNAc-4-sulfotransferase 2; N-acetylgalactosamine 4-O-sulfotransferase 2; N-acetylgalactosamine-4-O-sulfotransferase 2; UNQ2549/PRO6175
Gene Symbols CHST9
Host Species Rabbit
Immunogen Recombinant Human Carbohydrate sulfotransferase 9 protein (33-129AA).
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 71367, 83539
Target Species Human, Mouse
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.