Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CHTF18 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5109543
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5109543 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5109543 Supplier Invitrogen™ Supplier No. PA5109543
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: GTMLAYSLTY RQERTPDGQY IYRLEPNVEE LCRFPELPAR KPLTYQTKQL IAREIEVEKM RRAEASARVE NSPQVDGSPP GLEGLLGGIG EKGVHRPAPR NHEQRLEHIM RRAAREEQPE KDFFGRVVVR STAVPSAGDT APEQDSVERR MGTAVGRSEV WFRFNEGVSN AVRRSLYIRD LL.

CHTF18, CHTF8.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CHTF18
Applications ELISA, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 2.02 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene Chtf18
Gene Accession No. Q8WVB6
Gene Alias 6030457M03Rik; C16orf41; C321D2.2; C321D2.3; C321D2.3 (novel protein); C321D2.4; C321D2.4 (novel protein); Chl12; Chl12 homolog; chromosome transmission fidelity factor 18; chromosome transmission fidelity protein 18 homolog; chromosome transmission fidelity protein 18 homolog; LOW QUALITY PROTEIN: chromosome transmission fidelity protein 18 homolog; Chtf18; Ctf18; CTF18, chromosome transmission fidelity factor 18; CTF18, chromosome transmission fidelity factor 18 homolog; CTF18, chromosome transmission fidelity factor 18 homolog (S. cerevisiae); hCTF18; homolog of yeast CHL12; RUVBL; some homology with holliday junction DNA helicase RUVB like; zgc:113153
Gene Symbols Chtf18
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 794-975 of human CHTF18 (NP_071375.1).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 287146, 63922
Target Species Human, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.