Learn More
Innovative Research Inc Chx X Prtn L Ply Aff Pur Lyo
Supplier:  Innovative Research Inc ICHAPROLAP1MG
Encompass_Preferred
Chicken Anti Protein L Polyclonal Affinity Purified Lyophilized 1mg - This polyclonal antibody from Innovative Research is in a Lyophilized format, buffered in Phosphate buffered saline, pH 7.2, 0.1% sodium azide. Chickens were immunized with purified recombinant protein L. After multiple immunizations, eggs were collected and dilapidated. Antibodies were immunoaffinity purified off protein L agarose. Lyophilized material should be kept at 4C until rehydrated and will remain stable for a minimum of 2 years. Rehydrate by adding 1.0 mL of reagent quality water to the product vial. Store the liquid solution at 4C for up to a week; thereafter, aliquots should be prepared and stored frozen. Stable for a minimum of 1 year at -20C. Optimal working dilutions should be determined for the particular assay conditions. This product can be used for many research applications, including ELISA 0.1 - 0.01 ug/ml, Lateral Flow, WB 0.1 - 0.01 ug/ml.
Catalog No. 50-274-4615
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.