Learn More
Invitrogen™ CHX10 Polyclonal Antibody

Description
A suggested positive control for this product is rat or mouse retinal tissue lysate.
Specifications
Specifications
| Antigen | CHX10 |
| Applications | Immunohistochemistry (Frozen), Western Blot |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.08% sodium azide |
| Gene | VSX2 |
| Gene Accession No. | P58304, Q61412, Q9IAL1 |
| Gene Alias | C. elegans ceh-10 homeo domain containing homolog; ceh-10 homeo domain containing homolog; ceh-10 homeodomain containing homolog; ceh-10 homeodomain-containing homolog; CHX10; Homeobox protein CHX10; HOX10; Hox-10; MCOP2; MCOPCB3; ocular retardation; or; RET1; Visual system homeobox 2; Vsx2 |
| Gene Symbols | VSX2 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.