Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Chymotrypsin Monoclonal Antibody (CHYMO10-104.10), Invitrogen™

Catalog No. PIMA546015 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
1 mg
100 μg
500 μg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA546015 1 mg
PIMA546156 100 μg
01-671-577 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. PIMA546015 Supplier Invitrogen™ Supplier No. MA546015
  • $1,187.00
    $1,068.65 / Each of 1

    Save $118.35

    10% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Mouse Monoclonal Antibody

MA1-10624 detects Chymotrypsin from human samples. MA1-10624 has been successfully used in ELISA and Western blot applications. The MA1-10624 immunogen is purified human pancreatic Chymotrypsin.

This gene encodes a member of the peptidase S1 family. The encoded protein is a serum calcium-decreasing factor that has chymotrypsin-like protease activity. Alternatively spliced transcript variants have been observed, but their full-length nature has not been determined.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Chymotrypsin
Applications ELISA
Classification Monoclonal
Clone CHYMO10-104.10
Concentration 2.0 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative; pH 7.4
Gene CTRC
Gene Accession No. Q99895
Gene Alias 1810044E12Rik; caldecrin; chymotrypsin C; chymotrypsin C (caldecrin); chymotrypsin-C; CLCR; Ctrc; ELA4; elastase 4; elastase IV; preprocaldecrin; serum calcium decreasing factor; serum calcium-decreasing factor
Gene Symbols CTRC
Host Species Mouse
Immunogen Purified human pancreatic Chymotrypsin.
Purification Method Protein A
Quantity 1 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11330
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG3
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.