Learn More
CPC Scientific Suc-Leu-Leu-Val-Tyr-AMC 25MG
SDP
Supplier:  CPC Scientific SUBS012C
Encompass
Fluorogenic peptide used in the analysis of chymotrypsin-like activity of trypsins in addition to chymase from rat mast cells. Furthermore, it is a substrate for a chymotrpsin-like enzyme from sperm extract of the ascidian Halocynthia roretzi and a strong inhibitor of fertilization. It is also a substrate for carboxypeptidase Y, proteinase yscE, porcine calpain isozymes I and II, papain, and the multicatalytic proteinase, ingensin.SEQUENCE: Suc-Leu-Leu-Val-Tyr-AMCONE-LETTER SEQUENCE: Suc-LLVY-AMCMOLECULAR FORMULA: C40H53N5O10MOLECULAR WEIGHT: 763.89STORAGE CONDITIONS: -20 5 CCAS REGISTRY NUMBER: [94367-21-2]SYNONYMS: Suc-Leu-Leu-Val-Tyr-AMCRESEARCH AREA: ApoptosisREFERENCES:SKU(s): SUBS-012A, SUBS-012B, SUBS-012C, SUBS-012D
Catalog No. 50-211-0484
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.