Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ CIA 1
GREENER_CHOICESDP

Catalog No. 749450
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
74-945-0 50 mg
74-941-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 74-945-0 Supplier Tocris Bioscience™ Supplier No. 7494/50

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

COUP-TFII inhibitor

CIA 1 is an inhibitor of the nuclear receptor COUP-TFII (IC50 ranged from 1.2 μM to 7.6 μM, in prostate cancer cell lines). In vitro, CIA 1 inhibits growth of multiple prostate cancer cell lines and suppresses COUP-TFII activity to regulate its target genes. In vivo, CIA 1 reduces tumor growth in prostate cancer xenograft mouse models.

Specifications

Chemical Name or Material N-[2-(3,4-Dimethoxyphenyl)ethyl]-6-methylthieno[2,3-d]pyrimidin-4-amine hydrochloride
CAS 1049691-47-5
Quantity 50 mg
Target Additional Nuclear Receptor Compounds
Molecular Formula C17H19N3O2S.HCl
Purity 0.98
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.