Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ cIAP2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA551700
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA551700 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA551700 Supplier Invitrogen™ Supplier No. PA551700
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: QHAKWFPRCE YLIRIKGQEF IRQVQASYPH LLEQLLSTSD SPGDENAESS IIHFEPGEDH SEDAIMMNTP VINAAVEMGF SRSLVKQTVQ RKILATGENY RLVNDLVLDL LNAEDEIREE ERERATEE Highest antigen sequence identity to the following orthologs: Mouse - 75%, Rat - 74%.

The protein encoded by this gene is a member of a family of proteins that inhibits apoptosis by binding to tumor necrosis factor receptor-associated factors TRAF1 and TRAF2, probably by interfering with activation of ICE-like proteases. The encoded protein inhibits apoptosis induced by serum deprivation but does not affect apoptosis resulting from exposure to menadione, a potent inducer of free radicals. The amino acid sequence predicts three baculovirus IAP repeat domains and a ring finger domain. Transcript variants encoding the same isoform have been identified.
TRUSTED_SUSTAINABILITY

Specifications

Antigen cIAP2
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.20 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene BIRC3
Gene Accession No. Q13489
Gene Alias AIP 2; AIP1; Api1; Api2; apoptosis inhibitor 1; apoptosis inhibitor 2; AW107670; baculoviral IAP repeat containing 3; baculoviral IAP repeat-containing 2; baculoviral IAP repeat-containing 3; baculoviral IAP repeat-containing protein 3; Birc2; Birc3; Cellular inhibitor of apoptosis 2; cIAP1; cIAP-1; cIAP2; C-IAP2; cIAP-2; H HIAP1; H MALT2; HAIP1; HIAP1; hIAP-1; HIAP2; IAP homolog; IAP homolog C; IAP1; IAP-1; IAP2; Inhibitor of apoptosis protein 1; MALT2; mammalian IAP homolog C; MIAP1; mIAP-1; MIAP2; MIHB; MIHC; RING finger protein 49; RING-type E3 ubiquitin transferase BIRC3; RNF49; TNFR2-TRAF signaling complex protein; TNFR2-TRAF-signaling complex protein 1
Gene Symbols BIRC3
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human cIAP2. Recombinant protein control fragment (Product #RP-101848).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 330
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.