Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CIAPIN1 Monoclonal Antibody (OTI1H4), TrueMAB™, OriGene
SDP

Catalog No. 501682335
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-168-2335 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-168-2335 Supplier OriGene Supplier No. TA808732
May include imposed supplier surcharges.
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

CIAPIN1 is a cytokine-induced inhibitor of apoptosis with no relation to apoptosis regulatory molecules of the BCL2 or CASP families. Expression of CIAPIN1 is dependent on growth factor stimulation.

Specifications

Antigen CIAPIN1
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone OTI1H4
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene CIAPIN1
Gene Accession No. Q6FI81
Gene Alias Anamorsin, DRE2, PRO0915
Gene Symbols CIAPIN1
Host Species Mouse
Immunogen Human recombit protein fragment corresponding to amino acids 97-312 of human CIAPIN1 produced in E.coli.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 57019
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.