Learn More
Abcam Cibacron Blue 3G-A, Anthraquinone Derivative Dye, Solid, 1 g
Description
- Molecular Weight: 774.2 Da
- Form: Solid
- Properties: Anthraquinone derivative dye and potent nucleotide-mimetic ligand. Ortho-isomer of Reactive Blue 2.
- Biological Activity: Inhibits Na+/K+-ATPase and antagonizes P2 receptors (IC50 values are 17.4 and 9.1 μM for P2Y and P2X, respectively).
- Composition: 100% 2-Anthracenesulfonic acid, 1-amino-4-[[4-[[4-chloro-6-[(2-sulfophenyl)amino]-1,3,5-triazin-2-yl]amino]-3-sulfophenyl]amino]-9,10-dihydro-9,10-dioxo
The product is subject to the following: Abcam Restricted Use Statement
Specifications
Specifications
| Content And Storage | Store at ambient temperature. |
| Form | Solid |
| Shipping Condition | Ambient |
| Substrate Properties | Chemical Substrate |
| Target Molecule | Na+/K+-ATPase, P2 Receptors (P2Y, P2X) |
| Quantity | 1 g |
| For Use With (Application) | Sample Prep and Detection |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.