Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CIC Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA583721
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA583721 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA583721 Supplier Invitrogen™ Supplier No. PA583721
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: PKRKMRRRSS CSSEPNTPKS AKCEGDIFTF DRTGTEAEDV LGELEYDKVP YSSLRRTLDQ RRALVMQLFQ DHGFFPSAQA TAAFQARYAD IFPSKVCLQL KIREVRQKIM QA Highest antigen sequence indentity to the following orthologs: Mouse - 98%, Rat - 98%.

This gene encodes a core component of the exosome. The mammalian exosome is required for rapid degradation of AU rich element-containing RNAs but not for poly(A) shortening. The association of this protein with the exosome is mediated by protein-protein interactions with ribosomal RNA-processing protein 42 and ribosomal RNA-processing protein 46.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CIC
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry, Western Blot
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene CIC
Gene Accession No. Q96RK0
Gene Alias 1200010B10Rik; Capicua; capicua homolog; capicua homolog (Drosophila); capicua transcriptional repressor; CIC; KIAA0306; mKIAA0306; Protein capicua homolog; protein capicua homolog; LOW QUALITY PROTEIN: protein capicua homolog
Gene Symbols CIC
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human CIC. Recombinant protein control fragment (Product #RP-102474).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23152
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.