Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ CID 1375606
GREENER_CHOICE

Catalog No. 65445
Change view
Click to view available options
Quantity:
5 mg
25 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
65-445 5 mg
65-442-5 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 65-445 Supplier Tocris Bioscience™ Supplier No. 6544/5
Add to Cart
Add to Cart
This item is not returnable. View return policy

GPR27 agonist

CID 1375606 is a GPR27 agonist. Promotes recruitment of β-arrestin 2 to chimeric GPR27V2 and wild-type GPR27 in a firefly luciferase complementation assay (pEC50 values are 6.34 and 5.1, respectively).

Specifications

Chemical Name or Material 2,4-Dichloro-N-(phenylcarbamoyl)phenylbenzamide
CAS 313493-80-0
Quantity 5 mg
Molecular Formula C20H14Cl2N2O2
Purity 98%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.