Learn More
Cid1 Poly (U) Polymerase, 60 U, G-Biosciences
SCI_ED
Catalog No. S001133
Change view
Click to view available options
Promoter:
T7
Catalog No. Promoter
S001133 T7
1 options
Catalog No. S001133 Supplier G-Biosciences Supplier No. 7862269
Add to Cart
Add to Cart
Cid1 Poly (U) Polymerase is a template-independent RNA polymerase that catalyzes the addition of uridine monophosphates (UMP) to the 3′ end of RNA using UTP as a substrate.
  • Poly(U) and Poly(A) tailing of RNA for cloning
  • Labeling of RNA with UTP
Specifications
Polymerase Poly(A) RNA Polymerase
Product Line Ambion
Promoter T7