Learn More
Santa Cruz Biotechnology Anti-CIITA Antibody (7-1H), 200 ug/mL
Supplier:  Santa Cruz Biotechnology SC13556
Encompass
Anti-CIITA Antibody (7-1H) is recommended for detection of CIITA of mouse, rat and human origin by WB, IP, IF and IHC(P).
Catalog No. NC0706696
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.