Learn More
TARGETMOL CHEMICALS INC PF-06840003 25MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T430725MG
Encompass_Preferred
Also available in 2 mg 5 mg 10 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. PF-06840003 (EOS200271) is a specific and oral active IDO-1 inhibitor. purity: 99%
Catalog No. 50-257-6781