Learn More
Medchemexpress LLC HY-A0166 50mg , Cilastatin CAS:82009-34-5 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYA016650MG
Encompass_Preferred
HY-A0166 50mg Cilastatin CAS: 82009-34-5 Cilastatin (MK0791) is a reversible, competitive renal dehydropeptidase I inhibitor with an IC50 of 0.1 μM. Cilastatin inhibits the bacterial metallob-lactamase enzyme CphA with an IC50 of 178 μM. Cilastatin is an antibacterial adjunct. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-226-0277
Only null left
Add to Cart
This item is not returnable. View return policy
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.