Learn More
Selleck Chemical LLC Cilofexor
SDP
Supplier:  Selleck Chemical LLC S65475MG
Encompass_Preferred
Cilofexor(GS-9674) is a potent selective oral agonist of farnesoid X receptor (FXR) with an EC50 of 43 nM It has demonstrated anti-inflammatory antifibrotic effects and reduced portal pressure in preclinical liver fibrosis models showing potential therapeutic benefit for primary sclerosing cholangitis (PSC) and nonalcoholic steatohepatitis (NASH) research
Catalog No. 50-000-02819
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.