Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abcam Cimaterol, Beta Adrenoceptor Agonist, 99% Purity, Solid, 50 mg

Catalog No. 502707093
Encompass_Preferred
Change view
Click to view available options
Quantity:
50 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-270-7093 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-270-7093 Supplier Abcam Supplier No. AB12095450MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Potent β adrenoceptor agonist with >99% purity, active in vivo.

  • Activity: β adrenoceptor agonist (Ki = 2.32 μM); active in vivo.
  • Purity: >99%
  • Molecular Weight: 219.28 Da
  • Form: Solid
  • CAS Number: 54239-37-1
  • Storage: Store at ambient temperature.

The product is subject to the following: Abcam Restricted Use Statement

Specifications

Target Molecule Beta Adrenoceptor
Shipping Condition Ambient
Content And Storage Store at ambient temperature.
Form Solid
Formulation 100% Cimaterol
Quantity 50 mg
For Use With (Application) Beta adrenoceptor agonist
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.