Learn More
Abcam Cimaterol, Beta Adrenoceptor Agonist, Solid, >99% Purity, 10 mg
Description
- Molecular weight: 219.28 Da
- Purity: >99%
- Potency: β adrenoceptor agonist (Ki = 2.32 μM)
- Activity: Active in vivo
- Form: Solid
- Storage: Store at ambient temperature
The product is subject to the following: Abcam Restricted Use Statement
Specifications
Specifications
| Target Molecule | Beta Adrenoceptor |
| Substrate Properties | Chemical Substrate |
| Content And Storage | Store at ambient temperature. |
| Shipping Condition | Ambient |
| Buffer | 100% Cimaterol |
| Form | Solid |
| Formulation | 100% Cimaterol |
| Quantity | 10 mg |
| For Use With (Application) | Sample Prep and Detection |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.