Learn More
Cayman Chemical CImaterol-d7 5mg
SDP
Supplier:  Cayman Chemical 289035 MG
Encompass_Preferred
An internal standard for the quantification of cimaterol by GC- or LC-MS
Catalog No. 50-293-8769