Learn More
Sartorius CIMmultus OH-1 mL, 2 μm, Monolithic Column, Hydroxyl chemistry, 1 mL volume, 2 μm channel size, Non-cGMP Compliant, Operating flow rate: 0.1-16 mL/min
Catalog No. NC3397772 Shop All Sartorius Products
Encompass
CIMmultus OH-1 mL, 2 μm, Monolithic Column, Hydroxyl chemistry, 1 mL volume, 2 μm channel size, Non-cGMP Compliant, Operating flow rate: 0.1-16 mL/min
Catalog No. NC3397772 Supplier Sartorius Supplier No. 311.81402
CIMmultus OH-1 mL, 2 μm, Monolithic Column, Hydroxyl chemistry, 1 mL volume, 2 μm channel size, Non-cGMP Compliant, Operating flow rate: 0.1-16 mL/min