Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Rat CINC-1 (CXCL1) ELISA Kit

Catalog No. 502545379
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 x 96 Tests
5 x 96 Tests
96 Tests
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-254-5379 5 x 96 Tests
ERCXCL1 96 Tests
50-254-5380 10 x 96 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 50-254-5379 Supplier Invitrogen™ Supplier No. ERCXCL1X5
Add to Cart
Add to Cart

ELISA

The Rat CINC-1 (CXCL1) ELISA quantitates Rt CINC-1 in rat serum, plasma, or cell culture medium. The assay will exclusively recognize both natural and recombinant Rt CINC-1. Principle of the method The Rat CINC-1 solid-phase sandwich ELISA (enzyme-linked immunosorbent assay) is designed to measure the amount of the target bound between a matched antibody pair. A target-specific antibody has been pre-coated in the wells of the supplied microplate. Samples, standards, or controls are then added into these wells and bind to the immobilized (capture) antibody. The sandwich is formed by the addition of the second (detector) antibody, a substrate solution is added that reacts with the enzyme-antibody-target complex to produce measurable signal. The intensity of this signal is directly proportional to the concentration of target present in the original specimen. Rigorous validation Each manufactured lot of this ELISA kit is quality tested for criteria such as sensitivity, specificity, precision, and lot-to-lot consistency. See manual for more information on validation.

CXCL1, CXCL2, and CXCL3, also known respectively as GRO alpha, GRO beta (MIP-2 alpha) and GRO gamma (MIP-2 beta), are members of the CXC subfamily of chemokines. Mature CXCL1/2/3 proteins bind with high affinity to the IL-8 receptor type B and are potent neutrophil attractants and activators. In vitro, the processed forms GRO-alpha(4-73), GRO-alpha(5-73) and GRO-alpha(6-73) show a 30-fold higher chemotactic activity.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P14095
Assay Range 234.4 to 15,000 pg/mL
Assay Sensitivity 50 pg/ml
Calibration Range 300 pg/mL to 15000 pg/mL
Description CINC-1 (CXCL1) ELISA Kit, 96 Assays, 1 Kit, Rat Species, Chemokines and Receptors Protein Family, Immunology Research Discipline, Enzyme-Linked Immunosorbent Assay Designed for the Quantitation and Detection of CINC-1 (CXCL1) in Samples, Using a Microplat
Detectable Analytes CINC-1 (CXCL1)
Assay Solid-phase sandwich ELISA
Conjugate HRP
Detection Method Colorimetric
Product Type ELISA
Protein Family Chemokine & Receptors
Sample Type Plasma, Serum, Supernatant
Specificity This ELISA kit shows no cross-reactivity with any of the cytokines tested: rat CINC-2, CINC-3, CNTF, IL-1 alpha, IL-1 beta, IL-6, IL- 10, GM-CSF, IFN-gamma, Leptin, Lix, MCP-1, MIP-3 alpha, beta-NGF, TIMP-1, TNF-alpha, VEGF.
Substrate Type TMB
Sufficient For 96 assays
For Use With (Application) ELISA, Protein Assays, Protein Biology, Cancer Biology, Immunology, Neuroscience, Neurobiology, Protein Detection, Protein Analysis
For Use With (Equipment) Colorimetric Microplate Reader
Gene ID (Entrez) 81503
Gene Symbol CINC-1,Gro1
Host Species Rat
Interassay CV <12%
Intraassay CV <10
Kit Contents Pre-coated 96 well plate, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers
Label or Dye HRP
Quantity 5 x 96 Tests
Regulatory Status RUO
Research Discipline Immunology
Sample Volume Plasma 50 μL, Serum 50 μL, Supernatant 100 μL
Storage Requirements 2°C to 8°C
Synonym CINC-1, Gro1
Target CXCL1 (GRO alpha)
Test Time 1 hrs 20 mins
Total Assay Time 4 hrs 45 mins
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.