Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cinchonidine 98.0+%, TCI America™
SDP

Catalog No. C0347100G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C0347100G 100 g
C034725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C0347100G Supplier TCI America Supplier No. C0347100G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 485-71-2
Molecular Formula C19H22N2O
Molecular Weight (g/mol) 294.40
MDL Number MFCD00006783
InChI Key KMPWYEUPVWOPIM-UHFFFAOYNA-N
Synonym cinchonidine
PubChem CID 45358337
IUPAC Name {5-ethenyl-1-azabicyclo[2.2.2]octan-2-yl}(quinolin-4-yl)methanol
SMILES OC(C1CC2CCN1CC2C=C)C1=C2C=CC=CC2=NC=C1

Specifications

Melting Point 206°C
Color White-Yellow
Quantity 100 g
Formula Weight 294.40
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Cinchonidine
EINECSNumber (9)-0501
RTECSNumber GD2975000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.