Learn More
Cincinnati Surgical Sterile-Stainless Steel Blades and Scalpels
Description
- Made from British stainless steel
- 200% visual inspection
- Narrow grinding angle resulting in increased initial keenness
- Medium surface finish
- Strong foil package
- Handles are made of plastic (sold separately)
Specifications
Specifications
| Blade Material | British Stainless Steel |
| Product Type | Blade |
| Size | # 20 |
| Description | Medium Surface Finish |
| Sterility | Sterile |
| Quantity | 10/Pk. |