Learn More
Cincinnati Surgical Swann Morton Non-Sterile Carbon Steel Blades
SDP
Catalog No. 19200250
Change view
Click to view available options
Size:
# 10A
# 60
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-200-250 # 10A
19-200-251 # 60
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 19-200-250 Supplier Cincinnati Surgical Supplier No. 02SM10A
For use in wide variety of medical applications, including Dermaplaning
  • 200% visual inspection
  • Packaged with VCI Paper to protect blade from corrosion
  • Strongest/smallest foil package available
  • Narrow grinding angle resulting in increased initial keenness
  • High surface finish
  • Cellophane overwrap
  • Extended Range
Specifications
Blade Material Carbon Steel
Product Type Blade
Size # 10A
Description High Surface Finish
Sterility Non-sterile