Learn More
Cincinnati Surgical Lance Sterile Carbon Blades
Description
- 100% visual inspection
- Wide grinding angle for high durability
- Low surface finish
Certifications
CE Marked
Specifications
Specifications
| Blade Material | British Cold-rolled Surgical Carbon Steel |
| Size | # 15 |
| Description | Low Surface Finish |
| Sterility | Sterile |