Learn More
Medchemexpress LLC HY-N0610A 100mg Medchemexpress, Cinnamic acid CAS:621-82-9 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYN0610A100MG
Encompass_Preferred
Medchemexpress, HY-N0610A 100mg Cinnamic acid CAS:621-82-9 Cinnamic acid has potential use in cancer intervention, with IC50s of 1-4.5 mM in glioblastoma, melanoma, prostate and lung carcinoma cells. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-2805
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.