Learn More
Selleck Chemical LLC NLRP3 Inflammasome Inhibitor I S3680-1g
SDP
Supplier:  Selleck Chemical LLC S36801G
Encompass_Preferred
NLRP3 Inflammasome Inhibitor I an intermediate substrate in the synthesis of glyburide which is free of the cyclohexylurea moiety involved in insulin release is a novel inhibitor of the NLRP3 inflammasome
Catalog No. 50-310-8754
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.