Learn More
Medchemexpress LLC Cinnamic acid-d6 | 91453-04-2 | MFCD01863724 | 99.0% | 154.20 g·mol⁻¹ | C9H2D6O2 | 10 MG

Supplier: Medchemexpress LLC HYN0610AS10MG
Cinnamic acid-d6 is the deuterium-labeled form of cinnamic acid used as a stable isotope standard and tracer in analytical, mass spectrometry, and biochemical applications. It is reported to show activity in certain cancer cell lines (IC50 ~1-4.5 mM).
- Deuterium-labeled cinnamic acid for isotope tracing and analytical standards.
- High purity 99.0%.
- Molecular weight 154.20 g·mol⁻¹.
- CAS number 91453-04-2.
- Supplied as 10 mg powder.
- Recommended storage: powder at -20°C (3 years) or 4°C (2 years); in solvent at -80°C (6 months) or -20°C (1 month).
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.