Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cinnamoyl Chloride 97.0+%, TCI America™
SDP

Catalog No. P0133500G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P0133500G 500 g
P013325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P0133500G Supplier TCI America Supplier No. P0133500G

Chemical Identifiers

CAS 17082-09-6
Molecular Formula C9H7ClO
Molecular Weight (g/mol) 166.60
MDL Number MFCD00000732
InChI Key WOGITNXCNOTRLK-VOTSOKGWSA-N
Synonym cinnamoyl chloride, cinnamic acid chloride, cinnamic chloride, trans-3-phenylacryloyl chloride, 2-propenoyl chloride, 3-phenyl, cinnamoylchloride, e-cinnamoyl chloride, trans-cinnamoyl chloride, 2e-3-phenylprop-2-enoyl chloride, 3-phenyl-2-propenoyl chloride
PubChem CID 5354261
IUPAC Name (2E)-3-phenylprop-2-enoyl chloride
SMILES ClC(=O)\C=C\C1=CC=CC=C1

Specifications

Color White-Yellow
Quantity 500 g
UN Number 3261
Formula Weight 166.60
Percent Purity ≥97.0% (T)
Physical Form Crystalline Lumps
Chemical Name or Material Cinnamoyl Chloride
EINECSNumber (3)-1720
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.