Learn More
Medchemexpress LLC HY-77641 10mg Medchemexpress, Cinnamoylglycine CAS:16534-24-0 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY7764110MG
Encompass_Preferred
Medchemexpress, HY-77641 10mg Cinnamoylglycine CAS:16534-24-0 Cinnamoylglycine is a glycine conjugate of cinnamic acid and a urinary metabolite in human. Cinnamoylglycine is used as a potential urinary biomarker indicating intact or disrupted colonization resistance during and after antibiotic treatment. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-203-0981
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.