Learn More
Selleck Chemical LLC Flavokawain A
SDP
Supplier:  Selleck Chemical LLC S56005MG
Encompass_Preferred
Flavokawain A extracted from kava is an apoptotic inducers and anticarcinogenic agent Flavokawain A can down-regulation of antiapoptotic proteins such as XIAP survivin and Bcl-xL thereby changing the balance between apoptotic and antiapoptotic molecules and then induce cell death in tumor cells
Catalog No. 50-000-03156
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.