Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CINP Monoclonal Antibody (OTI5A11), TrueMAB™, OriGene
SDP

Catalog No. 501684502
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-168-4502 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-168-4502 Supplier OriGene Supplier No. TA812289
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The function of this gene has not been reported. This gene contains a trinucleotide (ctt) repeat length polymorphism; alleles with either one or two repeat units have been identified.

Specifications

Antigen CINP
Applications Western Blot
Classification Monoclonal
Clone OTI5A11
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene CINP
Gene Accession No. Q9BW66
Gene Alias cyclin dependent kinase 2 interacting protein
Gene Symbols CINP
Host Species Mouse
Immunogen Full length human recombit protein of human CINP produced in HEK293T cell.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51550
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.