Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

STERIS CIP 100™ Alkaline Process and Research Cleaner

Catalog No. 17355271 Shop All STERIS Corporation Products
Change view
Click to view available options
Volume (English):
1 gal.
5 gal.
30 gal.
55 gal.
200 gal.
250 gal.
300 gal.
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (English)
17-355-271 250 gal.
50-011-72 1 gal.
17-463-024 5 gal.
17-355-272 30 gal.
17-463-004 55 gal.
17-355-270 200 gal.
17-463-022 300 gal.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Catalog No. 17-355-271 Supplier STERIS Supplier No. 1D10D7
Add to Cart
Add to Cart
Request a Quote

Specially formulated to meet the unique cleaning demands found in industries that require an effective, validatable detergent.

CIP 100 Alkaline Process and Research Cleaneris specially formulated to meet the unique cleaning demands found in the pharmaceutical, biotechnology, cosmetic, dietary supplement, and other industries requiring an effective validatable detergent.

HOW CIP 100 WORKS

CIP 100 Detergent is a proprietary blend of potassium hydroxide, an advanced surfactant system and other performance-enhancing ingredients that provide multiple cleaning mechanisms. This low foaming product removes a wide range of process residues, from fermentation by-products to silicone-based emulsions and lubricants, and is ideal for use in clean-in-place (CIP), clean-out-of-place (COP) and manual applications.

WHY CIP 100?

  • Flexibility - Effective cleaning against a wide variety of pharmaceutical process residues in clean-in-place (CIP), clean-out-of-place (COP), and manual applications.
  • Easy to Validate - Facilitates cleaning validation with a comprehensive support documentation package including specific and non-specific analytical methods, toxicity, and substrate compatibility reports.
  • Cost Savings - Reduces water consumption and cleaning time due to superior rinsing and low foaming characteristics.
  • Global Availability - and manufactured with biodegradable surfactants to support harmonization of cleaning processes.
  • Reliability - Manufactured for the pharmaceutical industry in a cGMP compliant, FDA registered and ISO 9001 certified facility

Specifications

Product Type Alkaline Cleaner
Color-coded No
Form Clear Liquid
pH 12.4 (1% Solution)
Scent Unscented
Sterility Non-sterile
Size 250 gal.
Volume (English) 250 gal.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.