Learn More
Description
- A proprietary blend of phosphoric acid, an advanced surfactant system and other performance-enhancing ingredients that provide multiple cleaning mechanisms
- Low foaming detergent is ideal for use in CIP, COP, and manual applications for a variety of applications including derouging and passivation
- Facilitates cleaning validation with a comprehensive support documentation package including specific and non-specific analytical methods, toxicity, and substrate compatibility reports
- Reduces water consumption and cleaning time due to superior rinsing and low foaming characteristics
- Manufactured with biodegradable surfactants to support harmonization of cleaning processes
- Manufactured for the pharmaceutical industry in a cGMP compliant, FDA registered and ISO 9001 certified facility
Specifications
Specifications
| Product Type | Disinfectant |
| pH | 1.8 to 2.2 |
| Sterility | Non-sterile |
| Appearance | Clear |
| Composition | Phosphoric Acid, Citric Acid |
| Container Type | Tote |
| For Use With (Application) | CIP, COP, Rouge Removal, and Passivation of Process Equipment |
| Solubility | Complete |
| Volume (English) | 250 gal. |
Not yet available in the State of California
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.