Learn More
Cayman Chemical Ciprofloxacin (hydrochloride), 93107-08-5, 5 g
Supplier: Cayman Chemical 142865
Molecular Formula C17H18FN3O3 • HCl, Purity ≥95%, Formula Weight 367.8, Synonyms: 1-cyclopropyl-6-fluoro-1,4-dihydro-4-oxo-7-(1-piperazinyl)-3-quinolinecarboxylic acid, monohydrochloride, BAY-o 9867, NSC 620634; A fluoroquinolone antibiotic; active against a variety of Gram-positive and Gram-negative bacteria in vitro, including S. aureus, L. monocytogenes, P. aeruginosa, Legionella, N. gonorrhoeae, and H. pylori (MIC50s = 0.004-1 g/ml); active against clinical isolates of Bacteroides, Fusobacterium, Eubacterium, Actinomyces, Peptococcus, Peptostreptococcus, and Streptococcus in vitro (MIC50s = 0.5-2 g/ml); inhibits S. aureus DNA gyrase and topoisomerase IV (IC50s = 13.5 and 5.76 g/ml, respectively); reduces mortality in mouse models of intraperitoneal E. coli, P. vulgaris, K. pneumoniae, P. aeruginosa, and S. aureus infection (ED90-100s = 1-5, 2.5-5, 5-10, 20-40, and 80 mg/kg, respectively); prevents mortality in a mouse model of subcutaneous S. typhimurium infection at 10 mg/kg.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.