Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ciprofloxacin 98.0+%, TCI America™
SDP

Catalog No. C251025G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C251025G 25 g
C25105G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C251025G Supplier TCI America Supplier No. C251025G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 85721-33-1
Molecular Formula C17H18FN3O3
Molecular Weight (g/mol) 331.347
MDL Number MFCD00185755
InChI Key MYSWGUAQZAJSOK-UHFFFAOYSA-N
Synonym ciprofloxacin, ciprofloxacine, ciprobay, ciproxan, ciprofloxacina, cipro, ciprofloxacino, ciprofloxacinum, cipro iv, bacquinor
PubChem CID 2764
ChEBI CHEBI:100241
IUPAC Name 1-cyclopropyl-6-fluoro-4-oxo-7-piperazin-1-ylquinoline-3-carboxylic acid
SMILES C1CC1N2C=C(C(=O)C3=CC(=C(C=C32)N4CCNCC4)F)C(=O)O

Specifications

Melting Point 265°C
Color White
Quantity 25 g
Formula Weight 331.35
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Ciprofloxacin
RTECSNumber VB1993800
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.