Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CIQ, Tocris Bioscience™
Click to view available options
Quantity:
10 mg
50 mg
Description
Subunit-selective potentiator of NMDA receptors containing the GluN2C (formally NR2C) or GluN2D (formally NR2D) subunit. Increases channel opening efficiency and enhances NMDA receptor responses.
Please refer to IUPHAR Guide to Pharmacology for the most recent naming conventions.
Specifications
Quantity | 10 mg |
Synonym | ciq, 3-chlorophenyl 6,7-dimethoxy-1-4-methoxyphenoxy methyl-3,4-dihydroisoquinolin-2 1h-yl methanone, 3-chlorophenyl 3,4-dihydro-6,7-dimethoxy-1-4-methoxyphenoxy methyl-2 1h-isoquinolinyl methanone, 3-chlorophenyl 6,7-dimethoxy-1-4-methoxyphenoxy methyl-3,4-dihydroisoquinolin-2 1 h-yl methanone, 3-chlorophenyl-1r-6,7-dimethoxy-1-4-methoxyphenoxy methyl-3,4-dihydro-1h-isoquinolin-2-yl me, 3-chlorophenyl-6,7-dimethoxy-1-4-methoxyphenoxy methyl-3,4-dihydro-1h-isoquinolin-2-yl methanone, 3-chlorophenyl 3,4-dihydro-6,7-dimethoxy-1-4-methoxyphenoxy methyl-2 1h-isoquinolinyl-methanone, 2-3-chlorobenzoyl-6,7-dimethoxy-1-4-methoxyphenoxy methyl-1,2,3,4-tetrahydroisoquinoline |
Formula Weight | 467.94 |
Percent Purity | >98% |
Chemical Name or Material | CIQ |
Safety and Handling
Recommended Storage | Store at +4°C |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction