Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CIRBP Recombinant Rabbit Monoclonal Antibody (3V9S7)
GREENER_CHOICE

Catalog No. PIMA537911
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA537911 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA537911 Supplier Invitrogen™ Supplier No. MA537911
Only null left
Add to Cart
Add to Cart

Rabbit Recombinant Monoclonal Antibody

Positive Samples: 293T, HepG2, SH-SY5Y Immunogen sequence: MASDEGKLFV GGLSFDTNEQ SLEQVFSKYG QISEVVVVKD RETQRSRGFG FVTFENIDDA KDAMMAMNGK SVDGRQIRVD QAGKSSDNRS RGYRGGSAGG RGFFRGGRGR GRGFSRGGGD RGYGGNRFES RSGGYGGSRD YYSSRSQSGG YSDRSSGGSY RDSYDSYATH NE.

CIRBP is cold-inducible mRNA binding protein that plays a protective role in the genotoxic stress response by stabilizing transcripts of genes involved in cell survival. It acts as a translational activator and seems to play an essential role in cold-induced suppression of cell proliferation. It binds specifically to the 3'-untranslated regions (3'-UTRs) of stress-responsive transcripts RPA2 and TXN. This protein acts as a translational repressor and promotes assembly of stress granules (SGs), when overexpressed.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CIRBP
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Recombinant Monoclonal
Clone 3V9S7
Concentration 0.8 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.05% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Gene CIRBP
Gene Accession No. P60824, P60825, Q14011
Gene Alias A18 hnRNP; A18HNRNP; CIRBP; CIRP; cold inducible RNA binding protein; cold inducible RNA-binding protein; cold-inducible RNA-binding protein; glycine-rich RNA binding protein; glycine-rich RNA-binding protein CIRP; R74941; testicular tissue protein Li 39
Gene Symbols CIRBP
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-172 of human CIRBP (Q14011).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1153, 12696, 81825
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.