Learn More
Description
- Circle mode counting grid
- Available for 90 mm or 150 mm Petri dishes
Specifications
Specifications
| Product Type | Counting Grids |
| Display Type | None |
| Diameter (Metric) | 90 mm |
| For Use With (Equipment) | 90 mm Petri Dishes |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
