Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ Circlegrow™ Agar

Catalog No. 113100132 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
908 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-310-0132 908 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-310-0132 Supplier MP Biomedicals™ Supplier No. 113100132
Add to Cart
Add to Cart

Circlegrow™ Agar is designed for rapid growth of E. coli and high yields of plasmid DNA.

  • CIRCLEGROW™ Agar media is used for growth of E.coli.
  • It also extends plasmid production past log phase.
  • Acceptable pH range is 6.9 ± 0.3 at 25°C.
  • Please note that the pH of CIRCLEGROW™ Medium liquid increases as the temperature of the solution decreases.
TRUSTED_SUSTAINABILITY

Specifications

Concentration 55 g/L
Form Powder
Grade Molecular Biology Certified™
Media Type Circlegrow
Preparation Method Reagents are tested and chosen for their ability to promote optimum growth of E. coli when combined in Circlegrow™ Agar formulations. Chosen reagents are combined in the accurate proportions according to the proprietary Circlegrow™ Agar formula and thoroughly blended for a uniform distribution.
Product Type Nutrient Agar
Formulation Proprietary formulation containing Yeast Extract, Tryptone Broth, and Vitamins. Biological buffers have been added for proper pH adjustment.
For Use With (Application) Cell Culture, Proteomics, Purification and Detection, Recombinant Protein Expression and Analysis, Bacterial Culture, Plasmid Production, Protein Expression, Gene Expression, Cloning, Genetic Manipulation
Quantity 908 g
Solubility Soluble in water, 55 g/L
Synonym Enhanced Bacterial Media
Show More Show Less

Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.