Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ CIRCLEGROW™ Capsules, Molecular Biology Certified

Catalog No. p-4474870 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
1000 Capsules
250 Capsules
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP113000121 250 Capsules
MP113000131 1000 Capsules
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MP113000121 Supplier MP Biomedicals™ Supplier No. 113000121
Only null left
Add to Cart
Add to Cart

Designed for rapid growth of E. coli and high yields of plasmid DNA.

  • Encapsulated growth media for growth of E.coli
  • Extends plasmid production past log phase/li>
  • Enhanced bacterial media
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage 15°C to 30°C
Format Capsule
Grade Molecular biology grade
Product Type Microbiology Media
Formulation Yeast extract, Tryptone broth, Vitamins, Biological Buffers have been added for proper pH adjustment
For Use With (Application) For growth of Escherichia coli and certified for plasmid and other bacterial cultures in liquid medium
Packaging Type Container
Quantity 250 Capsules
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.