Learn More
Fine Science Tools CIRCULAR SAW FOR MICRO DRILL
Supplier: Fine Science Tools 1800020
Product: Circular Saw for Micro Drill/ Diameter: 1.6cm/ Thickness: 0.1mm/ Alloy/Material: Stainless Steel