Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mopec™ Circular Specimen Chucks for Leica, TBS, and Tanner Cryostats
SDP

Catalog No. 22445777 Shop All Mopec Products
Change view
Click to view available options
Color:
Blue
Green
Orange
Pink
Purple
Red
White
Yellow
Length (Metric):
22 mm
25 mm
30 mm
40 mm
55 mm
33 product options available for selection
Product selection table with 33 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color Length (Metric)
22-445-777 Purple 30 mm
22-445-492 Blue 22 mm
22-445-797 Green 22 mm
22-445-321 Orange 22 mm
22-445-796 Pink 22 mm
22-445-487 Purple 22 mm
22-445-330 Yellow 22 mm
22-445-331 Blue 25 mm
22-445-613 Green 25 mm
22-445-604 Orange 25 mm
22-445-282 Pink 25 mm
22-445-281 Purple 25 mm
22-445-788 Red 25 mm
22-445-684 White 25 mm
22-445-776 Blue 30 mm
22-445-786 Green 30 mm
22-445-609 Orange 30 mm
22-445-283 Pink 30 mm
22-445-341 Red 30 mm
22-445-738 White 30 mm
22-445-712 Yellow 30 mm
22-445-748 Blue 40 mm
22-445-880 Green 40 mm
22-445-877 Orange 40 mm
22-445-581 Pink 40 mm
22-445-879 Purple 40 mm
22-445-806 Red 40 mm
22-445-545 White 40 mm
22-445-559 Yellow 40 mm
22-445-352 Blue 55 mm
22-445-687 Orange 55 mm
22-445-490 Pink 55 mm
22-445-254 Red 55 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 33 options available.
33 options
Catalog No. 22-445-777 Supplier Mopec™ Supplier No. SP103PR
Add to Cart
Add to Cart

Available in multiple sizes and colors for use with Leica, TBS, and Tanner Cryostats.

  • Round, aluminum specimen chucks
  • Suitable for Leica (except CM1950 model), TBS, and Tanner Cryostats

Specifications

For Use With (Equipment) Leica, TBS, and Tanner Cryostats
Length (Metric) 30 mm
Color Purple
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.