Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abcam Circulating DNA Quantification Kit, Fluorescent, 10 to 1,000 ng/mL, 48 Test

Catalog No. 502582825
Encompass_Preferred
Change view
Click to view available options
Quantity:
48 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-258-2825 48 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-258-2825 Supplier Abcam Supplier No. AB15689848TEST
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Rapidly quantify circulating DNA in serum or plasma in 30 minutes.

  • Assay time: 30 min
  • Sensitivity: ≥ 10 ng/mL
  • Detection method: Fluorescent
  • Range: 10 to 1,000 ng/mL
  • Sample types: Plasma and Serum
  • Results type: Quantitative
  • Instrument compatibility: Microplate reader

The product is subject to the following: Abcam Restricted Use Statement

Specifications

Run Time 30 min.
Detection Location In-Solution Detection
For Use With (Equipment) Microplate Reader
Format Kit
No. of Reactions 48 Tests
Target Molecule DNA
Detection Method Fluorescence
Sample Type Plasma, Serum
Quantity 48 Tests
For Use With (Application) DNA quantification
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.