Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abcam Circulating DNA Quantification Kit, Quantitative, Fluorescent, 96 Test

Catalog No. 502582824
Encompass_Preferred
Change view
Click to view available options
Quantity:
96 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-258-2824 96 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-258-2824 Supplier Abcam Supplier No. AB15689896TEST
May include imposed supplier surcharges.
Add to Cart
Add to Cart

Quantifies DNA in biofluids within 30 minutes using a fluorescent assay.

  • Detection method: Fluorescent
  • Assay time: 30 min
  • Sensitivity: ≥ 10 ng/mL
  • Range: 10 to 1,000 ng/mL
  • Sample types: Plasma, Serum
  • Results type: Quantitative
  • Instrument compatibility: Microplate reader

The product is subject to the following: Abcam Restricted Use Statement

Specifications

Target Molecule DNA
Run Time 30 min.
For Use With (Equipment) Microplate Reader
No. of Reactions 96 Tests
Format Kit
Detection Method Fluorescence
Sample Type Plasma, Serum
Quantity 96 Tests
For Use With (Application) DNA quantification
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.