Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

cis-1,2-Bis(diphenylphosphino)ethylene, 97%

Catalog No. AAA1685903
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1685903 1 g
AAA1685906 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1685903 Supplier Thermo Scientific Chemicals Supplier No. A1685903
Add to Cart

CAS: 983-80-2 | C26H22P2 | 396.41 g/mol

Intermediate in organic syntheses.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Intermediate in organic syntheses.

Solubility
Insoluble in water.

Notes
Air Sensitive. Store in a cool, dry, conditions in well sealed container. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 983-80-2
Molecular Formula C26H22P2
Molecular Weight (g/mol) 396.41
MDL Number MFCD00003046 MFCD00063219
InChI Key NCKJIJSEWKIXAT-DQRAZIAOSA-N
Synonym cis-1,2-bis diphenylphosphino ethylene, z-1,2-bis diphenylphosphino ethene, cis-1,2-bis diphenylphosphino ethene, cis-vinylenebis diphenylphosphine, trans-1,2-bis diphenylphosphinyl ethylene, trans-vinylenebis diphenylphosphine, z-2-diphenylphosphanyl ethenyl diphenylphosphane, dppv, phosphine, trans
PubChem CID 2733415
SMILES C(=C\P(C1=CC=CC=C1)C1=CC=CC=C1)\P(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 122°C to 126°C
Quantity 1 g
Beilstein 757792
Sensitivity Air Sensitive
Solubility Information Insoluble in water.
IUPAC Name [(1Z)-2-(diphenylphosphanyl)ethenyl]diphenylphosphane
Formula Weight 396.41
Percent Purity 97%
Chemical Name or Material cis-1,2-Bis(diphenylphosphino)ethylene
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 213-569-7
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.