Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

cis-1,2-Bis(phenylsulfonyl)ethylene 95.0+%, TCI America™
SDP

Catalog No. B15735G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B15735G 5 g
B15731G 1 g
B157325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. B15735G Supplier TCI America Supplier No. B15735G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 963-15-5
Molecular Formula C14H12O4S2
Molecular Weight (g/mol) 308.366
MDL Number MFCD00010140
InChI Key YGBXMKGCEHIWMO-QXMHVHEDSA-N
PubChem CID 5383944
IUPAC Name [(Z)-2-(benzenesulfonyl)ethenyl]sulfonylbenzene
SMILES C1=CC=C(C=C1)S(=O)(=O)C=CS(=O)(=O)C2=CC=CC=C2

Specifications

Melting Point 101°C
Color White-Yellow
Quantity 5 g
Formula Weight 308.37
Percent Purity ≥95.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material cis-1,2-Bis(phenylsulfonyl)ethylene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.